Skip to Content
Merck
All Photos(2)

Key Documents

405949

Sigma-Aldrich

1,3-Bis(isocyanatomethyl)cyclohexane

mixture of cis and trans, 99%

Synonym(s):

1,3-Bis(methylisocyanate)cyclohexane, Hydrogenated m -xylylene diisocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(CH2NCO)2
CAS Number:
Molecular Weight:
194.23
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

0.4 mmHg ( 98 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.485 (lit.)

density

1.101 g/mL at 25 °C (lit.)

SMILES string

O=C=NCC1CCCC(CN=C=O)C1

InChI

1S/C10H14N2O2/c13-7-11-5-9-2-1-3-10(4-9)6-12-8-14/h9-10H,1-6H2

InChI key

XSCLFFBWRKTMTE-UHFFFAOYSA-N

General description

1,3-Bis(isocyanatomethyl)cyclohexane is a highly reactive compound with two cyanate groups which allows the formation of tailored polymers with enhanced elasticity, thermal stability, and chemical resistance. It can form biodegradable hydrogels that facilitates its use in controlled drug delivery. It is also widely used as a cross linking agent to enhance the durability and stability of adhesives and coatings.

Application

1,3-Bis(isocyanatomethyl)cyclohexane can be used:
  • As a precursor to synthesize pressure sensitive adhesives with strong adhesive strength and rapid strain recovery. These adhesives with high optical transmittance can be used in foldable displays, medical devices, and wearable electronics.
  • As a cross linking agent to prepare gel polymer electrolyte for Li-ion batteries using UV curing.
  • To fabricate niche-mimicking polymer hydrogels to identify molecular targets for treatment of pancreatic cancer.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1C - Skin Sens. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Simpson et al.
American journal of industrial medicine, 30(1), 48-55 (1996-07-01)
Twenty-three of 34 workers who had worked in the injection molding operation making polyurethane foam parts at an automobile parts manufacturing plant developed respiratory symptoms and/or systemic symptoms over a 2-month period following the full production use of a new
Meishan Chen et al.
Molecules (Basel, Switzerland), 24(8) (2019-04-21)
In recent years, polyureas with dynamic hindered urea bonds (HUBs), a class of promising biomedical polymers, have attracted wide attention as a result of their controlled hydrolytic properties. The effect of the chemical structures on the properties of polyureas and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service