Skip to Content
Merck
All Photos(1)

Key Documents

370193

Sigma-Aldrich

Trichlorooxobis(triphenylphosphine)rhenium(V)

Synonym(s):

Bis(triphenylphosphine)rhenium oxytrichloride, Bis(triphenylphosphine)trichlororhenium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)3P]2ReOCl3
CAS Number:
Molecular Weight:
833.14
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: rhenium
reagent type: catalyst

mp

205 °C (dec.) (lit.)

SMILES string

Cl[Re](Cl)(Cl)=O.c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6

InChI

1S/2C18H15P.3ClH.O.Re/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;;;/h2*1-15H;3*1H;;/q;;;;;;+3/p-3

InChI key

VJYQERSQTMFWJE-UHFFFAOYSA-K

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service