Skip to Content
Merck
All Photos(2)

Key Documents

265012

Sigma-Aldrich

Tungsten(VI) oxychloride

98%

Synonym(s):

Tungsten(VI) chloride oxide, Tungsten(VI) oxide chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
WOCl4
CAS Number:
Molecular Weight:
341.65
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
98%
form:
solid
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

solid

reaction suitability

reagent type: catalyst
core: tungsten

mp

211 °C (lit.)

SMILES string

Cl[W](Cl)(Cl)(Cl)=O

InChI

1S/4ClH.O.W/h4*1H;;/q;;;;;+4/p-4

InChI key

LQNAFXBTXJHEAX-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

  • Tungsten and titanium oxide thin films obtained by the sol-gel process as electrodes in electrochromic devices: The study discusses the preparation of tungsten trioxide using tungsten(VI) oxychloride dissolved in isopropanol for electrochromic applications (C Rizzuto, RC Barberi, M Castriota, 2022).
  • Flux Crystal Growth of Lanthanide Tungsten Oxychlorides, La8.64W6O30.45Cl, Ce8.64W5.74O30Cl, and Ln8.33W6O30Cl (Ln = Pr, Nd): Structural Stability ...: Examines the structural stability of lanthanide and tungsten-based oxychloride compounds (D Carone, VV Klepov, MD Smith, 2019).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service