Skip to Content
Merck
All Photos(1)

Documents

14845

Sigma-Aldrich

1,3-Bis(3-glycidyloxypropyl)tetramethyldisiloxane

≥95.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H34O5Si2
CAS Number:
Molecular Weight:
362.61
Beilstein:
223966
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.452

bp

184 °C/2 mmHg (lit.)

density

0.996 g/mL at 20 °C (lit.)

SMILES string

C[Si](C)(CCCOCC1CO1)O[Si](C)(C)CCCOCC2CO2

InChI

1S/C16H34O5Si2/c1-22(2,9-5-7-17-11-15-13-19-15)21-23(3,4)10-6-8-18-12-16-14-20-16/h15-16H,5-14H2,1-4H3

InChI key

MFIBZDZRPYQXOM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,3-Bis(3-glycidyloxypropyl)tetramethyldisiloxane can be used:
  • As a starting material to synthesize functional poly(siloxane amine)s via an amine-epoxy click polymerization for material and biological applications.
  • To prepare polymer electrolyte membranes (PEM) for fuel cell applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel clickable and fluorescent poly (siloxane amine) s for reusable adhesives and reprocessable elastomers
Zhou J, et al.
Polym. Chem., 11, 4780-4786 (2020)
Interpenetrating network-forming sulfonated poly (vinyl alcohol) proton exchange membranes for direct methanol fuel cell applications
Tseng C-Y, et al.
International Journal of Hydrogen Energy, 36, 11936-11945 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service