MilliporeSigma
  • Effect of prostaglandin I2 analogs on macrophage inflammatory protein 1α in human monocytes via I prostanoid receptor and cyclic adenosine monophosphate.
Effect of prostaglandin I2 analogs on macrophage inflammatory protein 1α in human monocytes via I prostanoid receptor and cyclic adenosine monophosphate.
Journal of investigative medicine : the official publication of the American Federation for Clinical Research (2014-01-10)
Ming-Kai Tsai, Chong-Chao Hsieh, Hsuan-Fu Kuo, San-Nan Yang, Chang-Hung Kuo, Ming-Yii Huang, Ying-Ming Tsai, Min-Sheng Lee, Chih-Hsing Hung
RÉSUMÉ
Inflammation plays critical roles in atherosclerosis. Chemokines are responsible for leukocyte trafficking and involve in inflammatory diseases. Macrophage inflammatory protein 1α (MIP-1α) has been implicated in atherosclerotic lesion formation. Prostaglandin I2 (PGI2) analog, used in pulmonary hypertension, has been reported to have anti-inflammatory functions. However, little is known about its role in the MIP-1α production in human monocytes. We investigated the effects of 3 conventional (iloprost, beraprost, and treprostinil) and 1 new (ONO-1301) PGI2 analogs, on the expression of MIP-1α expression in human monocytes. Human primary monocytes from control subjects and THP-1 cell line were treated with PGI2 analogs, with or without lipopolysaccharide (LPS) stimulation. Supernatants were harvested to measure MIP-1α levels by enzyme-linked immunosorbent assay. To explore which receptors involved the effects of PGI2 analogs on the expression of MIP-1α expression, I prostanoid (IP) and E prostanoid, peroxisome proliferator-activated receptor (PPAR)-α, and PPAR-r receptor antagonists were used to pretreat THP-1 cells. Forskolin, a cyclic adenosine monophosphate (cAMP) activator, was also used to further confirm the cAMP involvement on the effect of PGI2 analogs in MIP-1α production. Three PGI2 analogs could suppress LPS-induced MIP-1α production in THP-1 cells and human primary monocytes. ONO-1301 had a similar effect. CAY 10449, an IP receptor antagonist, could reverse the suppressive effects on MIP-1α production of iloprost. Forskolin, a cAMP activator, also suppressed MIP-1α production in THP-1 cells. Prostaglandin I2 analogs suppressed LPS-induced MIP-1α production in human monocytes via the IP receptor and cAMP pathway. The PGI2 analog may be potential in the treatment for atherosclerosis.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Treprostinil, ≥98% (HPLC)