MilliporeSigma
Catalytic properties of a cobalt metal-organic framework with a zwitterionic ligand synthesized in situ.
Dalton transactions (Cambridge, England : 2003) (2017-10-31)
T A Grigolo, S D de Campos, F Manarin, G V Botteselle, P Brandão, A A Amaral, E A de Campos
RÉSUMÉ
Herein we describe the high yield synthesis of a highly crystalline cobalt(ii) MOF with a novel zwitterionic ligand made up of 3,3',4,4'-BPTC and 1,4-cyclohexanediamine, obtained in situ during the hydrothermal synthesis. The compound with the molecular formula C
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
trans-1,4-Diaminocyclohexane, ≥98.0% (GC)