MilliporeSigma
Lack of evidence for the direct activation of endothelial cells by adult female and microfilarial excretory-secretory products.
PloS one (2011-08-11)
Tiffany Weinkopff, Patrick Lammie
RÉSUMÉ
Lymphangiectasia (dilation of the lymphatic vessel (LV)) is pathognomonic for lymphatic filariasis. In both infected humans and animal models of infection, lymphangiectasia is not restricted to the site of the worm nest, but is found along the infected vessel. These observations argue that soluble products secreted by the worm could be mediating this effect by activating the lymphatic endothelial cells (LEC) lining the vessel. We tested the ability of filarial Excretory-Secretory products to activate LECs, but were unable to detect a direct effect of the Excretory-Secretory products on the activation of LEC as assessed by a variety of approaches including cellular proliferation, cell surface molecule expression and cytokine and growth factor production (although other mediators used as positive controls did induce these effects). Collectively, these results do not support the hypothesis that Excretory-Secretory products directly activate LECs.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anticorps anti-α-tubuline monoclonal antibody produced in mouse, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Anticorps de chèvre anti-IgG de souris, conjugué à la HRP, Upstate®, from goat