MilliporeSigma
  • Association of luteal blood flow with follicular size, serum estrogen and progesterone concentrations, and the inducibility of luteolysis by PGF
Association of luteal blood flow with follicular size, serum estrogen and progesterone concentrations, and the inducibility of luteolysis by PGF
Theriogenology (2016-09-28)
Semra Kaya, Cihan Kaçar, Bülent Polat, Armağan Çolak, Duygu Kaya, I Safa Gürcan, Heinrich Bollwein, Selim Aslan
RÉSUMÉ
The aim of this study was to investigate the compatibility of the visual evaluation result of the blood flow characteristics and the blood flow measurements of the CL and the predictability of the responses given by corpora lutea with varying levels of blood flow to an induction of luteolysis by a PGF
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
(R)-Pantetheine, ≥95.0% (HPLC)