MilliporeSigma
Pharmacokinetics and pharmacodynamics of the tetracyclines including glycylcyclines.
The Journal of antimicrobial chemotherapy (2006-07-04)
Kenneth N Agwuh, Alasdair MacGowan
RÉSUMÉ
The pharmacokinetics of tetracyclines and glycylcyclines are described in three groups. Group 1, the oldest group, represented by tetracycline, oxytetracycline, chlortetracycline, demeclocycline, lymecycline, methacycline and rolitetracycline is characterized by poor absorption after food. Group 2, represented by doxycycline and minocycline, is more reliably absorbed orally, while group 3, represented by the glycylcycline tigecycline, is injectable only, with an improved antibacterial spectrum compared with the tetracyclines. Though incompletely understood, the pharmacodynamic properties of the tetracyclines and glycylcyclines are summarized.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Chlortetracycline hydrochloride, ≥91.0% dry basis (HPLC)
Sigma-Aldrich
Chlortetracycline hydrochloride, BioReagent, suitable for fluorescence, from Streptomyces aureofaciens, ≥85.0% (HPLC)