MilliporeSigma
Lnk prevents inflammatory CD8⁺ T-cell proliferation and contributes to intestinal homeostasis.
European journal of immunology (2014-02-19)
Hiroko Katayama, Taizo Mori, Yoichi Seki, Masaki Anraku, Masanori Iseki, Masashi Ikutani, Yukiko Iwasaki, Nobuaki Yoshida, Kiyoshi Takatsu, Satoshi Takaki
RÉSUMÉ
The intracellular adaptor Lnk (also known as SH2B3) regulates cytokine signals that control lymphohematopoiesis, and Lnk(-/-) mice have expanded B-cell, megakaryocyte, and hematopoietic stem-cell populations. Moreover, mutations in the LNK gene are found in patients with myeloproliferative disease, whereas LNK polymorphisms have recently been associated with inflammatory and autoimmune diseases, including celiac disease. Here, we describe a previously unrecognized function of Lnk in the control of inflammatory CD8(+) T-cell proliferation and in intestinal homeostasis. Mature T cells from newly generated Lnk-Venus reporter mice had low but substantial expression of Lnk, whereas Lnk expression was downregulated during homeostatic T-cell proliferation under lymphopenic conditions. The numbers of CD44(hi) IFN-γ(+) CD8(+) effector or memory T cells were found to be increased in Lnk(-/-) mice, which also exhibited shortening of villi in the small intestine. Lnk(-/-) CD8(+) T cells survived longer in response to stimulation with IL-15 and proliferated even in nonlymphopenic hosts. Transfer of Lnk(-/-) CD8(+) T cells together with WT CD4(+) T cells into Rag2-deficient mice recapitulated a sign of villous abnormality. Our results reveal a link between Lnk and immune cell-mediated intestinal tissue destruction.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)