MilliporeSigma
  • Synthesis of enantiopure termini-differentiated heptane stereotriads. Application to side chain-functionalized tetrahydrofurans of IKD-8344.
Synthesis of enantiopure termini-differentiated heptane stereotriads. Application to side chain-functionalized tetrahydrofurans of IKD-8344.
Organic letters (2000-08-10)
W Jiang, D A Lantrip, P L Fuchs
RÉSUMÉ
Enantiopure epoxy cycloheptenyl sulfones syn-7b and anti-7b are prepared in five high-yielding and stereospecific operations from 1, 3-cycloheptadiene. These substrates serve as effective precursors for cis- and trans-substituted tetrahydrofurans (12, 10) which are segments of the antineoplastic agent IKD-8344.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1,3-Cycloheptadiene, 95%