MilliporeSigma
Dipyridamole for preventing stroke and other vascular events in patients with vascular disease.
The Cochrane database of systematic reviews (2007-07-20)
E L L M De Schryver, A Algra, J van Gijn
RÉSUMÉ
Patients with limited cerebral ischaemia of arterial origin are at risk of serious vascular events (4% to 11% annually). Aspirin reduces that risk by 13%. In one trial, adding dipyridamole to aspirin was associated with a 22% risk reduction compared with aspirin alone. However, a systematic review of all trials of antiplatelet agents by the Antithrombotic Trialists' Collaboration showed that, in high-risk patients, there was virtually no difference between the aspirin-dipyridamole combination and aspirin alone. To assess the efficacy and safety of dipyridamole versus control in the secondary prevention of vascular events in patients with vascular disease. We searched the Cochrane Stroke Group trials register (searched June 2006), the Cochrane Central Register of Controlled Trials (CENTRAL) (The Cochrane Library Issue 2, 2006), MEDLINE (1966 to May 2006) and EMBASE (1980 to May 2006). We contacted authors and pharmaceutical companies in the search for further data on published and unpublished studies. We selected randomised long-term secondary prevention trials with concealed treatment allocation, treatment for more than one month, starting within six months after presentation of an arterial vascular disease. Treatment consisted of dipyridamole with or without other antiplatelet drugs compared with no drug or an antiplatelet drug other than dipyridamole. Two review authors independently selected trials for inclusion, assessed trial quality and extracted data. Data were analysed according to the intention-to-treat principle. Twenty-nine trials were included, with 23019 participants, among whom 1503 vascular deaths and 3438 fatal and non-fatal vascular events occurred during follow up. Compared with control, dipyridamole had no clear effect on vascular death (relative risk (RR) 0.99, 95% confidence interval (CI) 0.87 to 1.12). This result was not influenced by the dose of dipyridamole or type of presenting vascular disease. Compared with control, dipyridamole appeared to reduce the risk of vascular events (RR 0.88, 95% CI 0.81 to 0.95). This effect was only statistically significant in patients presenting with cerebral ischaemia. For patients who presented with arterial vascular disease, there was no evidence that dipyridamole, in the presence or absence of another antiplatelet drug reduced the risk of vascular death, though it reduces the risk of further vascular events. This benefit was found only in patients presenting after cerebral ischaemia. There was no evidence that dipyridamole alone was more efficacious than aspirin.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Dipyridamole, ≥98% (HPLC)
Dipyridamole for peak identification, European Pharmacopoeia (EP) Reference Standard
Dipyridamole, European Pharmacopoeia (EP) Reference Standard