MilliporeSigma
  • Etretinate enhances the susceptibility of human skin squamous cell carcinoma cells to 5-aminolaevulic acid-based photodynamic therapy.
Etretinate enhances the susceptibility of human skin squamous cell carcinoma cells to 5-aminolaevulic acid-based photodynamic therapy.
Clinical and experimental dermatology (2008-12-17)
N Ishida, D Watanabe, Y Akita, A Nakano, N Yamashita, T Kuhara, T Yanagishita, T Takeo, Y Tamada, Y Matsumoto
RÉSUMÉ
Photodynamic therapy (PDT) with 5-aminolaevulinic acid (5-ALA) is a noninvasive and effective treatment for superficial skin cancers. Etretinate, a derivate of vitamin A, with the chemical formula ethyl(2E,4E,6E,8E)-9-(4-methoxy-2,3,6-trimethylphenyl)-3,7-dimethyl-2,4,6,8-nona-tetraenoate, has been reported to have antitumour effects and to regulate the proliferation and differentiation of skin cancers. In order to develop more efficient PDT, we investigated whether etretinate enhanced the cytotoxic action of ALA-based PDT against human squamous cell carcinoma cell line, HSC-5. The in vitro cytotoxicity was measured using the 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) assay. Apoptotic cells were detected by double-staining with fluorescent annexin V and propidium iodide. Intracellular protoporphyrin IX (PpIX) converted from exogenous ALA was measured by a fluorescence meter. HSC-5 cells pretreated with a nontoxic concentration of etretinate became more susceptible to the cytotoxic action of ALA-based PDT. Etretinate-pretreated cells underwent apoptosis in response to ALA-based PDT. Etretinate pretreatment resulted in enhanced accumulation of ALA-dependent intracellular PpIX. The results suggest that etretinate enhances the susceptibility of HSC-5 cells to ALA-based PDT via the intracellular increase of ALA-dependent PpIX. Etretinate might be useful for improvement of ALA-based PDT.
MATÉRIAUX
Référence du produit
Marque
Description du produit
USP
Acitretin Related Compound B, United States Pharmacopeia (USP) Reference Standard