MilliporeSigma
Antifungal activity of borrelidin produced by a Streptomyces strain isolated from soybean.
Journal of agricultural and food chemistry (2012-01-17)
Chong-Xi Liu, Ji Zhang, Xiang-Jing Wang, Ping-Ting Qian, Ji-Dong Wang, Ya-Mei Gao, Yi-Jun Yan, Shu-Zhen Zhang, Peng-Fei Xu, Wen-Bin Li, Wen-Sheng Xiang
RÉSUMÉ
In this study, an endophytic Streptomyces sp. neau-D50 with strong antifungal activity against Phytophthora sojae was isolated from healthy soybean root, using an in vitro screening technique. A bioactivity-guided approach was then employed to isolate and determine the chemical identity of bioactive constituents with antifungal activity from strain neau-D50. The structure of the antifungal metabolite was elucidated as borrelidin on the basis of spectral analysis. To our knowledge, this is the first report that borrelidin has strong antifungal activity against dominant race 1 of P. sojae with EC(50) and EC(95) of 0.0056 and 0.026 mg/L, respectively. The values were respectively 62.5- and 262.3-fold lower than those of the commercial fungicide metalaxyl, which has been used to treat soybean seed for the control of P. sojae . The in situ bioassays demonstrated that borrelidin at 10 mg/L reduced P. sojae race 1 lesions on soybean seedlings by 94.72% without affecting root growth. Thus, borrelidin might be a promising candidate for new antifungal agents against P. sojae.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Borrelidin, from Streptomyces parvulus, ≥98% (HPLC)