MilliporeSigma
  • Juvenile hormone III, hydroprene and a juvenogen as soldier caste differentiation regulators in three Reticulitermes species: potential of juvenile hormone analogues in termite control.
Juvenile hormone III, hydroprene and a juvenogen as soldier caste differentiation regulators in three Reticulitermes species: potential of juvenile hormone analogues in termite control.
Pest management science (2006-07-13)
Ivan Hrdý, Jelena Kuldová, Robert Hanus, Zdenek Wimmer
RÉSUMÉ
The efficacy of juvenile hormone III (JH III) and two JH-mimicking compounds was compared in laboratory experiments with Reticulitermes lucifugus (Rossi), R. santonensis Feytaud and R. virginicus (Banks). Induction of presoldier, soldier and/or soldier-worker intercaste differentiation was taken as positive response to the treatment. The novel juvenogen, a fatty acid ester of a juvenoid alcohol, induced greatest soldier differentiation in all species tested, followed by hydroprene. JH III was less effective. Representatives of three Reticulitermes species showed similar trends in soldier induction rates. Differences in mortality in treatments of termites of the same species from different colonies with the same compound were observed and evidently were caused by differences in the conditions of respective colonies.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
S-Hydroprene, PESTANAL®, analytical standard