MilliporeSigma
Action of ubenimex on aminopeptidase activities in spleen cells and peritoneal macrophages from mice.
The Journal of antibiotics (1987-11-01)
H Kuramochi, A Motegi, M Iwabuchi, K Takahashi, H Horinishi, H Umezawa
RÉSUMÉ
The action of ubenimex on aminopeptidase (APase) activity was studied in intact spleen cells and peritoneal macrophages from mice. Ubenimex strongly inhibited hydrolyzing activities against arginine-beta-naphtylamide (Arg-NA), Lys-NA and Pro-NA in both cells. Inhibition of hydrolysis of Leu-NA, Met-NA and Ala-NA was relatively small or not observed. When both cells were incubated in HANKS' solution, hydrolyzing activities against Arg-NA, Lys-NA and Pro-NA were released to the medium, while Leu-NA and Met-NA-hydrolyzing activities were mostly bound. In addition, the Leu-NA-hydrolyzing activity in the spleen cells was kinetically studied. The Arg-NA and Leu-NA-hydrolyzing activities in four fractions prepared from the homogenate of spleen cells were also studied kinetically. From these studies it was suggested that ubenimex inhibits aminopeptidase B and a Pro-NA-hydrolyzing enzyme more effectively than Leu-APase in intact spleen cells and peritoneal macrophages from mice.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
L-Arginine β-naphthylamide hydrochloride