MilliporeSigma
Nanocrystal Cu2O-loaded TiO2 nanotube array films as high-performance visible-light bactericidal photocatalyst.
Applied microbiology and biotechnology (2012-06-29)
Shengsen Zhang, Chang Liu, Xiaolu Liu, Haimin Zhang, Porun Liu, Shanqing Zhang, Feng Peng, Huijun Zhao
RÉSUMÉ
In this work, we report the use of a non-toxic nanocrystal Cu(2)O-loaded TiO(2) nanotube array (Cu(2)O/TNTs) film as high-performance visible-light bactericidal photocatalyst. The samples were characterized by field-emission scanning electron microscopy, X-ray photoelectron spectroscopy, and ultraviolet-visible diffusion reflection spectroscopy. This Cu(2)O/TNTs film photocatalyst is capable of complete inactivation of Escherichia coli in 5 × 10(7) colony-forming units/mL within a record short disinfection time of 20 min under visible-light irradiation. The average bactericidal percentage of the Cu(2)O/TNTs for E. coli under visible-light irradiation are 20 times and 6.6 times higher than those of TNTs under the same conditions and Cu(2)O/TNTs without light, respectively. This superior bactericidal performance is mainly attributed to the high ability to produce OH radicals by both photogenerated electron and hole of the prepared photocatalyst under visible light. The Cu(2)O/TNTs film photocatalyst makes it applicable to broad fields including drinking water disinfection.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Copper(I) oxide, powder, ≤7 μm, 97%
Sigma-Aldrich
Copper(I) oxide, ≥99.99% trace metals basis, anhydrous