MilliporeSigma
Composition of transgenic soybean seeds with higher γ-linolenic acid content is equivalent to that of conventional control.
Journal of agricultural and food chemistry (2012-02-14)
Fengyun Qin, Linzhi Kang, Liqiong Guo, Junfang Lin, Jingshen Song, Yinhua Zhao
RÉSUMÉ
γ-Linolenic acid (GLA) has been used as a general nutraceutical for pharmacologic applications, particularly in the treatment of skin conditions such as eczema. Four transgenic soybean lines that produce GLA at high yields (4.21% of total fatty acids, up to 1002-fold) were generated through the stable insertion of the Delta-6-fatty acid desaturase gene isolated from Borago officinalis into the genome of a conventional soybean cultivar. As part of the safety assessment of genetically engineered crops, the transgenic soybean seeds were compared with their parental soybean seeds (nontransgenic) by applying the principle of substantial equivalence. Compositional analyses were conducted by measuring the fatty acids, proximate analysis (moisture, crude protein, crude fat, carbohydrates, TDF, and ash contents), amino acids, lectins, and trypsin inhibitor activity. The present results showed that the specific transgenic cultivar studied was similar to the conventional control.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Acide γ-linolénique, ≥99%, liquid
Supelco
Acide γ-linolénique, analytical standard