MilliporeSigma
Extended para-hydrogenation monitored by NMR spectroscopy.
Chemical communications (Cambridge, England) (2010-11-17)
Joel A Tang, Francesca Gruppi, Roman Fleysher, Daniel K Sodickson, James W Canary, Alexej Jerschow
RÉSUMÉ
A system that provides a sustained hyperpolarized (1)H NMR signal in an aqueous medium is reported. The enhanced signal lasts much longer than typical (1)H T(1) values, uncovering new possibilities for implementing hyperpolarized (1)H NMR/MRI experiments or performing kinetics studies that would not otherwise be detectable.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Methyl 2-acetamidoacrylate, 98%