MilliporeSigma
  • Synthesis of some new anils: part 1. Reaction of 2-hydroxy-benzaldehyde and 2-hydroxynaphthaldehyde with 2-aminopyridene and 2-aminopyrazine.
Synthesis of some new anils: part 1. Reaction of 2-hydroxy-benzaldehyde and 2-hydroxynaphthaldehyde with 2-aminopyridene and 2-aminopyrazine.
Molecules (Basel, Switzerland) (2007-10-26)
Abdullah M Asiri, Khadija O Badahdah
RÉSUMÉ
New Schiff bases derived from 2-aminopyridene and 2-aminopyrazine have been synthesized. The UV-Visible spectra of the compounds have been investigated in acetonitrile and toluene. The compounds were in tautomeric equilibrium (enol-imine O- H...N, keto-amine O...H-N forms) in polar and nonpolar solvents. For some derivatives the keto-amine form was observed in both toluene and acetonitrile. 1H-NMR and IR results showed that all Schiff bases studied favor the enol-imine form over the keto form in a weakly polar solvent such as deuterochloroform.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Aminopyrazine, 98%