MilliporeSigma
A water-borne adhesive modeled after the sandcastle glue of P. californica.
Macromolecular bioscience (2008-12-02)
Hui Shao, Kent N Bachus, Russell J Stewart
RÉSUMÉ
Polyacrylate glue protein analogs of the glue secreted by Phragmatopoma californica, a marine polycheate, were synthesized with phosphate, primary amine, and catechol sidechains with molar ratios similar to the natural glue proteins. Aqueous mixtures of the mimetic polyelectrolytes condensed into liquid complex coacervates around neutral pH. Wet cortical bone specimens bonded with the coacervates, oxidatively crosslinked through catechol sidechains, had bond strengths nearly 40% of the strength of a commercial cyanoacrylate. The unique material properties of complex coacervates may be ideal for development of clinically useful adhesives and other biomaterials.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Sephadex® LH-20