MilliporeSigma
Label-free aptamer-based sensors for L-argininamide by using nucleic acid minor groove binding dyes.
Chemical communications (Cambridge, England) (2011-01-29)
Zece Zhu, Chuluo Yang, Xiang Zhou, Jingui Qin
RÉSUMÉ
The nucleic acid minor groove binding dyes, DAPI and Hoechst 33258, were for the first time used in label-free aptamer-based sensors for L-argininamide. The synergy binding effect results in the enhancement of fluorescence of dyes. The method for detection of L-argininamide is simple, rapid and cost-effective.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
L-Argininamide dihydrochloride