MilliporeSigma
Time course of blast-induced injury in the rat auditory cortex.
PloS one (2018-03-01)
Srinivasu Kallakuri, Edward Pace, Huichao Lu, Hao Luo, John Cavanaugh, Jinsheng Zhang
RÉSUMÉ
Blast exposure is an increasingly significant health hazard and can have a range of debilitating effects, including auditory dysfunction and traumatic brain injury. To assist in the development of effective treatments, a greater understanding of the mechanisms of blast-induced auditory damage and dysfunction, especially in the central nervous system, is critical. To elucidate this area, we subjected rats to a unilateral blast exposure at 22 psi, measured their auditory brainstem responses (ABRs), and histologically processed their brains at 1 day, 1 month, and 3-month survival time points. The left and right auditory cortices was assessed for astrocytic reactivity and axonal degenerative changes using glial fibrillary acidic protein immunoreactivity and a silver impregnation technique, respectively. Although only unilateral hearing loss was induced, astrocytosis was bilaterally elevated at 1 month post-blast exposure compared to shams, and showed a positive trend of elevation at 3 months post-blast. Axonal degeneration, on the other hand, appeared to be more robust at 1 day and 3 months post-blast. Interestingly, while ABR threshold shifts recovered by the 1 and 3-month time-points, a positive correlation was observed between rats' astrocyte counts at 1 month post-blast and their threshold shifts at 1 day post-blast. Taken together, our findings suggest that central auditory damage may have occurred due to biomechanical forces from the blast shockwave, and that different indicators/types of damage may manifest over different timelines.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Cocktail Mouse mAb (SMI-22), liquid, clone SMI-22, Calbiochem®