MilliporeSigma
  • Dasatinib plus Nutlin-3 shows synergistic antileukemic activity in both p53 wild-type and p53 mutated B chronic lymphocytic leukemias by inhibiting the Akt pathway.
Dasatinib plus Nutlin-3 shows synergistic antileukemic activity in both p53 wild-type and p53 mutated B chronic lymphocytic leukemias by inhibiting the Akt pathway.
Clinical cancer research : an official journal of the American Association for Cancer Research (2010-11-26)
Giorgio Zauli, Rebecca Voltan, Raffaella Bosco, Elisabetta Melloni, Sandra Marmiroli, Gian Matteo Rigolin, Antonio Cuneo, Paola Secchiero
RÉSUMÉ
To analyze the effect of the combination of Dasatinib, a multikinase inhibitor, plus Nutlin-3, a nongenotoxic activator of the p53 pathway, in primary B chronic lymphocytic leukemia (B-CLL) patient samples and B leukemic cell line models. The induction of cytotoxicity was evaluated in both primary B-CLL cell samples (n = 20) and in p53(wild-type) (EHEB, JVM-2) and p53(deleted/mutated) (MEC-2, BJAB) B leukemic cell lines. The role of Akt in modulating leukemic cell survival/apoptosis in response to Dasatinib or Dasatinib + Nutlin-3 was documented by functional experiments carried out using specific pharmacological inhibitors and by overexpression of membrane-targeted constitutively active form of Akt. The combination of Dasatinib + Nutlin-3 exhibited a synergistic cytotoxicity in the majority (19 out of 20) of B-CLL samples, including patients carrying 17p- (n = 4), and in both p53(wild-type) and p53(deleted/mutated) B leukemic cell lines. At the molecular level, Dasatinib significantly counteracted the Nutlin-3-mediated induction of the p53 transcriptional targets MDM2 and p21 observed in p53(wild-type) leukemic cells. Conversely, Nutlin-3 did not interfere with the ability of Dasatinib to decrease the phosphorylation levels of ERK1/2, p38/MAPK, and Akt in both p53(wild-type) and p53(deleted/mutated) B leukemic cell lines. A critical role of Akt downregulation in mediating the antileukemic activity of Dasatinib and Dasatinib + Nutlin-3 was demonstrated in experiments carried out by specifically modulating the Akt pathway. These findings suggest that Dasatinib + Nutlin-3 might represent an innovative therapeutic combination for both p53(wild-type) and p53(deleted/mutated) B-CLL.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-phospho-Akt1/PKBα (Ser473) Antibody, clone SK703, rabbit monoclonal, clone SK703, Upstate®, from rabbit
Sigma-Aldrich
Anti-ZAP-70 Antibody, clone 2F3.2, clone 2F3.2, Upstate®, from mouse