MilliporeSigma
Transfer of sulfamethazine from contaminated beeswax to honey.
Journal of agricultural and food chemistry (2010-05-27)
Wim Reybroeck, Frans J Jacobs, Hubert F De Brabander, Els Daeseleire
RÉSUMÉ
A liquid chromatographic tandem mass spectrometric method for the determination of sulfa drugs in beeswax was developed. When performing residue control on beeswax intended for the fabrication of wax foundations, residues of sulfonamides were found. A migration test was set up to study whether sulfonamide-containing beeswax could lead to the contamination of honey. The higher the concentration of sulfamethazine doped in the wax, the higher was the concentration of sulfamethazine found in the honey. The maximum transfer was 15.6, 56.9, and 29.5% of the initial amount spiked in the wax foundation. In a second experiment, the percentage of sulfamethazine migrating from medicated winter feed to beeswax in relation to the concentration in the syrup and the contact time was studied. The maximum transfer of sulfamethazine from medicated sucrose syrup to beeswax was 3.1%.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Amplifluor® UniPrimer II Fluorescein