MilliporeSigma
Serine/threonine protein phosphatases PP1 and PP2A are key players in apoptosis.
Biochimie (2003-10-31)
Alphonse Garcia, Xavier Cayla, Julien Guergnon, Frédéric Dessauge, Véronique Hospital, Maria Paz Rebollo, Aarne Fleischer, Angelita Rebollo
RÉSUMÉ
The reversible phosphorylation of proteins controlled by protein kinases and protein phosphatases is a major mechanism that regulates a wide variety of cellular processes. In contrast to C. elegans, recent studies in mammalian cells have highlighted a major role of serine/threonine protein phosphorylation in apoptosis. To illustrate the importance of dephosphorylation processes in apoptosis, this review will focus on recent studies suggesting that the interaction of the serine/threonine protein phosphatase 1 (PP1) and protein phosphatase 2A (PP2A) with certain regulators of the Bcl-2 family is critically involved in the control of apoptosis.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
PP1α Protein, 10 µg, Recombinant, full length, human PP1α, for use in Phosphatase Assays.