MilliporeSigma
The purification and subunit structure of a membrane-bound ATPase from the Archaebacterium Halobacterium saccharovorum.
Biochemical and biophysical research communications (1987-08-31)
L I Hochstein, H Kristjansson, W Altekar
RÉSUMÉ
A membrane-bound ATPase from Halobacterium saccharovorum was solubilized using sodium deoxycholate and Zwittergent 3-10 and purified by hydrophobic and ammonium sulfate-mediated chromatography. The enzyme, which had a molecular mass of 350 kDa, was composed of two major (87 and 60 kDa) and two minor (29 kDa and 20 kDa) subunits. The halobacterial ATPases appear to be unlike any other ATPase described to date.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3-(Decyldimethylammonio)­propane­sulfonate inner salt, zwitterionic detergent