MilliporeSigma
  • Stimulation of accumbal GABAB receptors inhibits delta1- and delta2-opioid receptor-mediated dopamine efflux in the nucleus accumbens of freely moving rats.
Stimulation of accumbal GABAB receptors inhibits delta1- and delta2-opioid receptor-mediated dopamine efflux in the nucleus accumbens of freely moving rats.
European journal of pharmacology (2018-08-08)
Yuriko Watanabe, Yuri Aono, Masamichi Komiya, John L Waddington, Tadashi Saigusa
RÉSUMÉ
The nucleus accumbens contains delta-opioid receptors that may decrease inhibitory neurotransmission. As GABAB receptors inhibit dopamine release, decrease in activation of GABAB receptors may be a mediator of delta-opioid receptor-induced accumbal dopamine efflux. If so, accumbal dopamine efflux induced by delta-opioid receptor activation should be suppressed by stimulating GABAB receptors. As delta-opioid receptors are further subdivided into delta1- and delta2-opioid receptors, we analysed the effects of the GABAB receptor agonist baclofen on delta1- and delta2-opioid receptor-mediated accumbal dopamine efflux in freely moving rats using in vivo microdialysis. Drugs were applied intracerebrally through the dialysis probe. Doses of compounds show total amount administered (mol) during 25-50 min infusions. Baclofen (2.5 and 5.0 nmol), which did not alter basal dopamine levels, inhibited the delta1-opioid receptor agonist DPDPE (5.0 nmol)-induced dopamine efflux. Baclofen (2.5 and 5.0 nmol) also inhibited the delta2-opioid receptor agonist deltorphin II (25.0 nmol)-induced dopamine efflux. A low dose of the GABAB receptor antagonist 2-hydroxysaclofen (100.0 pmol), which failed to alter basal accumbal dopamine levels, counteracted the inhibitory effects of baclofen (5.0 nmol) on DPDPE (5.0 nmol)- and deltorphin II (25.0 nmol)-induced dopamine efflux. The present results show that reduction in accumbal GABAB receptor-mediated inhibition of accumbal dopaminergic activity facilitates activation of delta1- and delta2-opioid receptor-induced increases in accumbal dopamine efflux. This study suggests that activation of delta1- and delta2-opioid receptors on the cell bodies and/or terminals of accumbal GABAergic interneurons inhibits GABA release and, accordingly, decreases GABAB receptor-mediated inhibition of dopaminergic terminals, resulting in enhanced accumbal dopamine efflux.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
R(+)-Baclofen hydrochloride, solid