MilliporeSigma
Système de purification d’eau Milli-Q® HX 7040 SD (LC) Centralized pure water solution for up to 800 L/day Type 2 water; for low chlorine feed water
Système de purification d’eau Milli-Q® HXSD
Système de purification d’eau Milli-Q® HX 7040 SD (LC) Centralized pure water solution for up to 800 L/day Type 2 water; for low chlorine feed water
Synonyme(s):
Système de purification d’eau Milli-Q® HXSD