PAMPO201
Profil`Air® Pump Module
About This Item
Code UNSPSC :
41104008
Nomenclature NACRES :
NB.24
Produits recommandés
Disponibilité
available only in EU
Paramètres
200 mL/min flow rate (calibrated)
Taille
20 × 10 × 10
Application(s)
environmental
industrial hygiene
Compatibilité
for sampling dynamic air flow
Description générale
This pump is calibrated to draw air at a flow rate of 200 mL/min. This is necessary for dynamic air sampling.
Coupled with the dynamic sampling module (PAMEX001-1EA), it creates a controlled air flow around the dynamic sampling sensors, thus accelerating the capture process of the material.
Coupled with the dynamic sampling module (PAMEX001-1EA), it creates a controlled air flow around the dynamic sampling sensors, thus accelerating the capture process of the material.
Adéquation
Fits the following instruments:
PAKSF421
PAMEX001
PAKSF421
PAMEX001
Autres remarques
Weight: 0.8
Temperature conditions: Room temperature
Temperature conditions: Room temperature
Informations légales
Profil'Air is a registered trademark of Ethera S.A.