Principaux documents
58257
Raccords Rheodyne®
long nut for 1/16 in. tubing, pkg of 10 ea
10 EA
110,00 $
110,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Changer de vue
10 EA
110,00 $
About This Item
Code UNSPSC :
41115700
110,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Matériaux
stainless steel nut
Description
long nut
Conditionnement
pkg of 10 ea
Raccords
long nut for 1/16 in. tubing
Description générale
The long Rheodyne nut has a longer hex portion for easier wrench access.
Produits recommandés
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Informations légales
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Les clients ont également consulté
Slide 1 of 1
1 of 1
Questions
  1. Hi. I have a few question about product#58257 (long nut Rheodyne Fittings). Do these come with ferrules? If they don't, can you suggest a ferrule that suits these nuts? What size is the hex nut?
    1 answer
    1. This product includes the nut only. Product 58258 is the 1/16” ferrule needed. The hex nut is 1/4 inch. Please see the link below to review the recommended ferrule product:
      https://www.sigmaaldrich.com/US/en/product/supelco/58258