Principaux documents
57627
Boucle d'injection pour injecteur modèle 7725
volume 100 μL
About This Item
Code UNSPSC :
41115854
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Volume
100 μL
Description générale
Sample loop for model 7725 injector (volume 100 μL) is best suited for advanced fluid-handling solutions for broad range of analytical instrumentation and in vitro diagnostic systems. It is useful in high performance liquid chromatographic (HPLC) systems and is quite accurate and precise in injecting 1 μL-5 mL concentration of samples.
Produits recommandés
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Articles
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Identifiez et corrigez les problèmes couramment rencontrés en chromatographie liquide haute performance (HPLC), afin de restaurer les performances de votre colonne.