Principaux documents
56625-U
Supelco
LC-MS Post Column Flow Splitters
Fixed, Split Ratio = 10:1
Synonyme(s) :
LC-MS Flow Divider
1 EA
3 390,00 $
3 390,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Changer de vue
1 EA
3 390,00 $
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
SB.52
3 390,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Description générale
LC-MS post column flow splitters with split ratio of 10:1 are ideally used for liquid chromatography with mass spectrometry and capillary chromatography. They generally use two compact fluid resistor elements which are designed as cartridges for easy replacement. They also provide a fixed split ratio with extremely low dead volume. And the split ratio is not affected by changes in solvent viscosity or pressure and is extremely stable and reproducible. Also used for post column applications.
Produits recommandés
Discover LiChropur reagents ideal for HPLC or LC-MS analysis