Principaux documents
56122
O-Ring pour extrémités de colonnes Omnifit®
Viton®, for use with 15 mm bore column
About This Item
Code UNSPSC :
12000000
Matériaux
Viton®
Conditionnement
pkg of 10 ea
Technique(s)
LPLC: suitable
Compatibilité
for use with 15 mm bore column
Description générale
O-ring sealing components are used in machines required to seal the fluids or gases.[1] Column end piece tend to trap liquid around the O-ring sealing, to avoid such a problem the upper column end piece can be redesigned.[2]
Informations légales
Omnifit is a registered trademark of Diba Industries
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Anthony Sofronas
Analytical Troubleshooting of Process Machinery and Pressure Vessels (2006)
Peptide synthesis. Part 13. Feedback control in solid phase synthesis. Use of fluorenylmethoxycarbonyl amino acid 3, 4-dihydro-4-oxo-1, 2, 3-benzotriazin-3-yl esters in a fully automated system.
Cameron, Linda R., et al.
Journal of the Chemical Society. Perkin Transactions 1, 10, 2895-2901 (1988)