28405-U
Adaptateur pour seringue Luer
I.D. 10 mm, pk of 3
Description
for processing Dual-Layer Carbon Reversible Tubes using a glass luer syringe
Conditionnement
pk of 3
Technique(s)
solid phase extraction (SPE): suitable
D.I.
10 mm
Application(s)
food and beverages
Choose from one of the most recent versions:
Certificats d'analyse (COA)