Principaux documents
24644
Supelco
Chargeur de batterie
for use with Model 1062 & 1063 Bag Samplers, AC/DC input 110 V AC
About This Item
Code UNSPSC :
41104008
Technique(s)
whole air sampling: suitable
Application(s)
air monitoring
environmental
industrial hygiene
Compatibilité
for use with Model 1062 & 1063 Bag Samplers
Description générale
Battery charger is a 110V AC charger used as accessories with a DC battery in a 10-liter vacuum bag sampler.