Principaux documents
23805
Supelco
Ecrou pour source MSD pour interface GC-MS
1/16 in. male hexagonal wrenchtight
About This Item
Code UNSPSC :
41115700
Conditionnement
pkg of 4 ea
Technique(s)
gas chromatography (GC): suitable
Raccords
1/16 in. male hexagonal wrenchtight
Description générale
Replacement nut for the Capillary Column Butt Connector.
Autres remarques
We offer a variety of chromatography accessories including analytical syringes