Principaux documents
21976-U
Coude mâle à 90°
Swagelok® 400-2-2, stainless steel, 1/8 in. male NPT, 1/4 in. Swagelok
Synonyme(s) :
RACCORD COUDÉ MÂLE EN ACIER INOXYDABLE SWAGELOK 400-2-2, 1/4 T x 1/8 P, UNITAIRE®
About This Item
Code UNSPSC :
41112500
eCl@ss :
32035302
Nomenclature NACRES :
SB.54
Matériaux
stainless steel
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Swagelok® 400-2-2
Raccords
1/4 in. Swagelok
1/8 in. male NPT
Description générale
Tube à male NPT. Ecrous et ferrules inclus.
Degree elbow is suitable to change direction of a pipe. In general, 45-degree or 90-degree elbows are used. Elbows enforce sudden inner diameter changes and rough edges which lead to turbulence and greater pressure drop. They are suitable for tubing systems. The capability to bend tubing delivers smoother transition.[1]
Informations légales
Swagelok is a registered trademark of Swagelok Company
Thwarting fluid-system leaks.
Cox, John C.
Chemical Processing (2002)