Principaux documents
Z762628
Eppendorf® Combitips advanced® pipette tips, Quality grade
maximum volume 0.1 mL
Synonyme(s) :
pipetting tips
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.01
Matériaux
white tip
Stérilité
non-sterile
Caractéristiques
barrier: no
Conditionnement
pack of 100 ea bag
Fabricant/nom de marque
Eppendorf® 0030089405
Volume maximum
0.1 mL
Description générale
The Combitips advanced are optimized for all Repeater models to create the perfect connection!
The new Eppendorf Combitips advanced feature the characteristic Eppendorf 3D design on the upper rim of the Combitip. With 9 volume sizes (0.1 mL to 50 mL) and Eppendorf Quality, Sterile, PCR Clean and Eppendorf Biopur purity grades, you will always find the perfect Combitip for your application.
With the redesigned dispenser box for the Eppendorf Quality tips, the extricable chute makes Combitips advanced always within reach. Portioning in 4 bags (25 pcs. each) ensures dust-free storage.
Autres remarques
Browse all pipette tips
Informations légales
Combitips advanced is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG