Principaux documents
Z757942
Sartorius Picus electronic pipette
8 channel, volume 50-1200 μL, pack of 1 ea
Synonyme(s) :
electronic pipetting, pipettors
1 EA
2 830,00 $
2 830,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
2 830,00 $
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
2 830,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Entrée CA/CC
100 V / 240 V AC, European 2-pin plug (flat 2-pin)
100 V / 240 V AC, UK plug (3 pin)
100 V / 240 V AC, US 2-pin plug
Caractéristiques
8-channel
CE compliant
electronic
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Sartorius 735391
Volume
50-1200 μL
Alimentation par piles
rechargeable
Adéquation
suitable for (Product is used in uHPLC, HPLC and trace analysis applications.)
Description générale
Sartorius Picus of the Biohitfamily is the smallest and lightest electronic pipette on the market helping to avoid pipetting caused symptoms like RSI (repetitive strain injury). The Picus handle design and the location of the operation buttons enables comfortable pipetting.
Electronic tip ejection, one hand-operated adjustment wheel, and conveniently located start button minimizes the force needed in pipetting. Repetitive and time consuming pipetting can be done accurately and fast with serial dispensing modes and a new Tracker - great help for example in microplate work.
  • Tracker in conjunction with pipetting, reverse pipetting and multi dispensing modes to help microplate work
  • Adjustment possibility for 1-point, 2-points and 3-points
  • Easy maintenance of lower parts
  • Safe-Cone filters in all models > 10 μl to prevent contaminations
  • Autoclavable lower parts in all models (excl. mcp 1200 μL)