Principaux documents
Z744066
EZcap FoilStrips
aluminum (chemically resistant), non-sterile
About This Item
Code UNSPSC :
41122410
Nomenclature NACRES :
NC.01
Matériaux
aluminum (chemically resistant)
Stérilité
non-sterile
Conditionnement
case of 300 ea
Fabricant/nom de marque
Excel Scientific F2-CS1X8-300
Catégories apparentées
Description générale
  • An economical and more user-friendly alternative to strip-caps; no more sore thumbs and fingers guaranteed!
  • Precision cut to a smaller dimension to seal a single 8-well row of a 96-well plate or a single PCR 8-tube strip
  • Each sheet contains 6 FilmStrips on a continuous backing liner for ease of use
  • Strips can be peeled individually or two-at-a-time for increased efficiency
  • Strips can be used to selectively cover one row of a plate to prevent accidental pipetting or to re-seal rows that have been pierced
  • Certified DNase- & RNase- free
  • 2.0 mil aluminum foil
  • Specially formulated adhesive provides an excellent seal to minimize evaporation, reduce cross-contamination and prevent spillage
  • FilmStrips remove easily and cleanly with no splash back as can occur with strip-caps
  • Features two removable end-tabs with perforations for ease of handling, application and removal
  • A non-locking workstation is recommended for sample prep (e.g. 96 round-well PCR plate or deep well block)
  • Removal of the end-tabs at the perforations is recommended before cycling
  • A final sealing step is highly recommended in the thermal cycler because 8-tube strips flex during handling
Informations légales
EZcap is a trademark of Excel Scientific, Inc.