Principaux documents
Z740464
MICROGARD 1500 PLUS coveralls for protection from particules and light spray
size M
1 EA
21,20 $
21,20 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
21,20 $
About This Item
Code UNSPSC :
46181503
Nomenclature NACRES :
NB.35
21,20 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Taille
M
Fabricant/nom de marque
Arco 1763104
Description générale
MICROGARD 1500 PLUS white coverall is made from a highly breathable anti-static SMS fabric which utilises the latest developments in micro-fibre technology to ensure good filtration efficiency, offers protection from particles, perfect for working with asbestos, powders and general direct and dust with added light liquid spray protection.
Features:
  • 3 piece hood
  • 2-way front zipper with re-sealable storm flap
  • Anti-static treatment
  • Elasticated hood, wrists, waist and ankles
  • Stitched seams
Approved to:
EN ISO 13982-1 Type 5 Particulate protection
EN13034 Type 6 Reduced Spray
EN1149-5 Anti-static
EN1073-2 Barrier to radioactive particulates - Class 1
Informations légales
Microgard is a trademark of Microgard Limited