Principaux documents
Z740361
Eppendorf® Reference® 2 Variable Volume Pipettor
volume - 0.1-2.5 μL, 2-20 μL (yellow), 20-200 μL, 100-1,000 μL, pack of 4 ea
Synonyme(s) :
Adjustable volume pipette
4 EA
2 360,00 $
2 360,00 $
En stockDétails
Changer de vue
4 EA
2 360,00 $
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
2 360,00 $
En stockDétails
Caractéristiques
electronic: no
single-channel
Conditionnement
pack of 4 ea
Fabricant/nom de marque
Eppendorf® 2231302003
Plage de mesure du volume
0.1-2.5 μL
100-1000 μL
2-20 μL
20-200 μL
Volume
1000 μL
Catégories apparentées
Description générale
The Reference 2 is the successor to the highly successful Reference pipette, and enhances the range′s already high standards with a new design, reduced weight and operating forces and multi-channel versions. As the premium product in Eppendorf′s pipette portfolio, the Reference 2 provides the most accurate results possible alongside robust, reliable handling and optimum user safety.
These levels of security make the Reference 2 ideal for use with precious liquids, or in any application requiring extremely high levels of accuracy. When there can be no compromise on quality and reproducability, Eppendorf′s new Reference 2 is an ideal instrument for these liquid handling applications.
Conditionnement
Includes: 4 adjustable-volume pipettes (0.1-2.5 μL, 2-20 μL (yellow), 20-200 μL, 100-1,000 μL), 1 full box of Eppendorf pipette tips for each pipette volume (excludes 5 mL and 10 mL tips)
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG