Principaux documents
Z740015
BRAND® filter tips, racked, TipRack
volume 0.5-10 μL, BIO-CERT®
960 EA
89,10 $
89,10 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
960 EA
89,10 $
About This Item
Code UNSPSC :
41121601
Nomenclature NACRES :
NB.02
Volume maximum:
1 μL
Volume:
0.5-10 μL
Stérilité:
sterile; autoclaved
Caractéristiques:
RNase and DNase free
endotoxin free
filter
BIO-CERT®
Conditionnement:
pack of 960 ea (10 racks of 96)
Fabricant/nom de marque:
BRAND 732624
89,10 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
colorless
colorless polypropylene tip and filter element (polyethylene)
Stérilité
sterile; autoclaved
Caractéristiques
RNase and DNase free
endotoxin free
filter
BIO-CERT®
Conditionnement
pack of 960 ea (10 racks of 96)
Fabricant/nom de marque
BRAND 732624
Volume maximum
1 μL
Volume
0.5-10 μL
Description générale
Pipette tips and filter tips are manufactured by BRAND in a cleanroom under the most modern production conditions, and are automatically rack packed and packaged to ensure that the tips are of consistently high quality.
Non-self-sealing filter tips from BRAND have a PE filter that is free from chemical additives. Permeability is controlled by the combination of pore size and filter length, so that no aerosols can reach the pipette shaft. These filters function with consistent reliability. On the other hand, liquids can pass very slowly should they accidentally contact the filter.
Since the filter does not swell as in the case of self-sealing filter tips, the sample can be recovered from the filter by simply actuating the pipette′s blow-out function, or by centrifugation if necessary. This is clearly an important advantage of non-self-sealing filters, especially when working with valuable samples. As an added advantage, the absence of filter additives protects samples from this source of contamination.
NEW! TipRack, non-sterile and sterile, BIO-CERT®
Space-saving refill unit for TipBox. Protected in an environmentally compatible packaging of recyclable PET. Compared to the filled TipBoxes, the amount of waste with the new refill units is reduced by 20%. All pipette tips and filter tips up to 1000 μL are free of DNA, RNases, endotoxins and ATP. TipRacks in BIO-CERT® quality are sterile according to ISO 11 137 and the AAMI guidelines, a SAL of 10-6 is obtained.
These racks are supplied with a transfer aid that enables simple, contamination-free transfer into a previously sterilized TipBox. All tip-trays are printed on one side with information on the contents.
Caractéristiques et avantages
• Cleanroom quality for contamination-free analyses.• Perfect fit for the Transferpette® micropipette and many other pipettes.• The tip system for all requirements.
Autres remarques
BR732624 (India)
Informations légales
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG