A graduated scale placed on the stage to be used as a standard of measurement for the specimen.
Z735620
Jenco™ compound microscopes
AC/DC input 115 V AC, brightfield, achromatic lens, monocular, mechanical stage; LED or other illumination
Synonyme(s) :
compound microscopes, microscopes
About This Item
Produits recommandés
Description
3-plug
Fabricant/nom de marque
Jenco CP135-M4
Disponibilité
not available in EU
Description générale
- Illumination can be a mirror, a rechargeable LED, an incandescent bulb or halogen bulb
- Eye pieces come in monocular heads, or teaching heads for assisting students in finding specimens and also binocular heads for easy viewing with both eyes
- Objectives are offered in multiple quality and quantity levels
- Video systems are available to project images on a screen
- Industry leading 5 year warranty
Choose from different frames for the right reason and teach science with the right equipment for your students.
Informations légales
Certificats d'analyse (COA)
Désolés, nous n'avons pas de COA pour ce produit disponible en ligne pour le moment.
Déjà en possession de ce produit ?
-
What is a Stage micrometer on a Jenco™ microscope?
1 answer-
Helpful?
-
-
Do Jenco™ inverted microscopes use oil objectives?
1 answer-
No, because the objective is below the slide and the drop of oil could only sit on top of the slide.
Helpful?
-
-
What Jenco™ microscopes can be used wiith video or digital camera equipment?
1 answer-
We recommend only those models with trinocular heads because they are more stable and we have adapters to fit the trinocular tube, not the eyepiece, Jenco Model BC-311 (Sigma nos. Z735086 for 115V or Z737062 for 220V).
Helpful?
-
-
Do microscopes just magnify things?
1 answer-
No, microscopes both magnify and clarify. They allow us to see small objects clearly.
Helpful?
-
-
Why use immersion oil on a Jenco™ microscope?
1 answer-
If an objective is designed to use oil, you must use oil to get an acceptable image. Oil provides an improved path for the image to travel between the specimen and the objective.
Helpful?
-
-
Do eyepieces add magnification for Jenco™ microscopes?
1 answer-
Yes. The standard eyepiece adds 10X magnification and is included in all models.
Helpful?
-
-
What is a LWD objective?
1 answer-
A Longer Working Distance for when you can't get close enough to focus on what you want to see. They are included with Jenco brand inverted microscopes.
Helpful?
-
-
What is a Reticle on a Jenco™ microscope?
1 answer-
A group of lines, shapes, circles, or angles that are inserted in the eyepiece (usually on a disc) that provide either a standard of measurement or a focal point for the observer.
Helpful?
-
-
What objectives can you use immersion oil with on a Jenco™ microscope?
1 answer-
Only those objectives designed for it with an oil specification clearly marked on it's barrel (100X OIL), otherwise the oil creeps up into the microscope causing problems.
Helpful?
-
-
What is a Stage or Mechanical stage on a Jenco™ microscope?
1 answer-
A flat plate for placing the slide/specimen with controls to easily move the sample both vertically and horizontally in small increments.