Principaux documents
Z719676
Sorenson low binding standard tips
maximum volume 1000 μL, non-sterile
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.01
Matériaux
colorless
polypropylene
Stérilité
non-sterile
Caractéristiques
barrier: no
Conditionnement
pack of 1000 ea (Bag)
case of 5000 ea (5 bags 1000 tips)
Fabricant/nom de marque
Sorenson 10200T
Volume maximum
1000 μL
Volume
1000 μL
Catégories apparentées
Description générale
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for research-grade pipettors.
Informations légales
Sorenson is a trademark of Sorenson BioScience, Inc.