Principaux documents
Z692743
microDOC Compact Gel Documentation System, with UV Transilluminator (UVT365)
AC/DC input 230 V AC
1 EA
10 250,00 $
10 250,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
10 250,00 $
About This Item
Code UNSPSC :
41105300
Nomenclature NACRES :
NB.22
10 250,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
Contains UK and Schuko plugs
Fabricant/nom de marque
Cleaver CSI-MDOCUV365
Description générale
System comprises of:
Canon G12 digital
2MB storage card
Ethidium bromide & close up +3 filters
Lift-off hood with easy access door and built-in inner lights
8 in. TFT color monitor
12 x 12 cm UV 365 nm transilluminator
This option combines the microDOC Compact Gel Documentation System with a 12 x 12 cm UV 365 nm tranilluminator.
Available with optional TotalLab 1D software. Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
Informations légales
TotalLab is a trademark of Nonlinear Dynamics Ltd.