Principaux documents
Z683183
Large Rodent Shipping Container
About This Item
Code UNSPSC :
41102601
Nomenclature NACRES :
NB.22
L × l × P
20 in. × 14.75 in. × 7 in., 295 total square inches. Configurable up to 4 compartments.
Application
Filtration Media:
  • Spunbond Polypropylene - 170 cfm/ft
Autoclave Cycles:
2 cycles maximum recommended up to 126ºC, 20 minutes maximum
  • Meets IATA (International Air Transport Association) specifications
  • All-plastic construction: box, lid, dividers and filter medium
  • Superior airflow via patented engineered openings
  • Easily stackable for material handling and transportation, with integral spacers to ensure proper air circulation
  • Clear plastic windows enables at-a-glance inspection of lab animals
  • Positive open/close lid facilitates animal insertion/removal
  • Interior rim provides a barrier against gang and go incidents
  • Manufactured in the USA
Chemical Resistance - Inert to most chemicals except:
  • 9Chloroform
  • Clorosulfonic Acid
  • Chlorine Gas
  • Carbon Tetrachloride
  • Ixooctane
  • Xylene
  • Nitric Acid
  • Petroleum Ether
  • Amyl chloride
  • Butyl Acetate
  • Benzene
  • Bromine Water
  • Most Petroleum fuels will attack
Caractéristiques et avantages
Premium Shipping Containers for Premium Research Models
  • Cardboard boxes may be okay for a batch of average lab rats, but SAGE research models are far from average. In most cases, you′re receiving professionally bred, highly validated animals. Many of them genetically engineered to your specifications
  • SAGE Labs offers plastic containers for shipping your SAGE research models more safely. These containers are sturdier than cardboard boxes and have excellent protection against contaminants, breakage, and leakage en route to your lab
  • The patented design also ensures unsurpassed convenience and ease of handling at every stage
Informations légales
Composants de kit seuls
Réf. du produit
Description
  • Container 1 ea
  • Lid 1 ea
  • Large Closure Label 1 ea
  • Small Closure Label 2 ea
  • Live Animal Label 2 ea
Composants de kit également disponibles séparément
Réf. du produit
Description
FDS
  • Large Rodent Box Divider 1 ea