Principaux documents
Z640409
Eppendorf® epT.I.P.S. Reloads
volume range 0.5-20 μL
Synonyme(s) :
pipetting tips
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.01
Matériaux
light gray tip
Stérilité
non-sterile
Caractéristiques
barrier: no
Conditionnement
pack of 960 (10 trays of 96 tips)
Fabricant/nom de marque
Eppendorf® 0030 073.041
Eppendorf® 022491521
Longueur
46 mm
Volume maximum
20 μL
Gamme de volumes
0.5-20 μL
Volume
0.5—20 μL
Description générale
Part of the ep T.I.P.S. box and reload system for contamination-free handling of refill trays, with plastic lids protecting the trays from contact during transfer to the working box.
Trays of tips which may be autoclaved in their packaging and stored clean for later use. Tips may be used straight from the reload tray, or loaded into ep T.I.P.S box (purchase separately)
Informations légales
Eppendorf is a registered trademark of Eppendorf AG