Principaux documents
Z640298
Eppendorf® epT.I.P.S. set
volume range 0.1-10 μL
Synonyme(s) :
pipetting tips
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.01
Matériaux
dark gray tip
Stérilité
non-sterile
Caractéristiques
barrier: no
Conditionnement
pack of 480 (5 trays of 96 tips, includes 1 reusable box)
Fabricant/nom de marque
Eppendorf® 0030 073.207
Eppendorf® 022491407
Longueur
34 mm
Volume maximum
10 μL
Gamme de volumes
0.1-10 μL
Volume
0.1—10 μL
Description générale
Part of the ep T.I.P.S. box and reload system for contamination-free handling of refill trays, with plastic lids protecting the trays from contact during transfer to the working box.
Autoclavable working box, optimized for multichannel pipettes, with removable lid and larger footprint to avoid tipping, supplied with 5 trays of tips.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG